1-Chloromethylpyrene ≥97.0% (通过滴定分析)

供应商: TCI EUROPE
Warning

TCIAC1637-1GEA 0 CNY
TCIAC1637-1G
1-Chloromethylpyrene ≥97.0% (通过滴定分析)
1-Chloromethylpyrene
分子式: C₁₇H₁₁Cl
分子量: 250,73 g/mol
存储条件: Freezer
MDL编号: MFCD00962113
CAS号: 1086-00-6

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (Argentmetric Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR