2-Ethoxyethyl cyanoacetate ≥98.0% (通过GC)

供应商: TCI EUROPE
Warning

TCIAC1756-500GEA 0 CNY
TCIAC1756-500G
2-Ethoxyethyl cyanoacetate ≥98.0% (通过GC)
2-Ethoxyethyl cyanoacetate
分子式: C₇H₁₁NO₃
分子量: 157,17 g/mol
闪点: 144°C
MDL编号: MFCD00071546
CAS号: 32804-77-6
EINECS: 251-228-4

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR