1-(2-Cyanoethyl)-2-methylimidazole ≥98.0% (通过GC,滴定分析)
供应商: TCI EUROPE
|
Danger
|
TCIAC1963-25GEA
0
CNY
TCIAC1963-25G
1-(2-Cyanoethyl)-2-methylimidazole ≥98.0% (通过GC,滴定分析)
1-(2-Cyanoethyl)-2-methylimidazole
分子式:
C₇H₉N₃ 分子量: 135,17 g/mol |
MDL编号:
MFCD00047049 CAS号: 23996-55-6 EINECS: 245-976-0 UN号: 3439 ADR: 6.1,III |
产品技术规格
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 55.0 to 59.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...