1-(2-Cyanoethyl)-2-phenylimidazole ≥98.0% (通过GC,滴定分析)

供应商: TCI EUROPE
Warning

TCIAC1964-25GEA 0 CNY
TCIAC1964-25G
1-(2-Cyanoethyl)-2-phenylimidazole ≥98.0% (通过GC,滴定分析)
1-(2-Cyanoethyl)-2-phenylimidazole
分子式: C₁₂H₁₁N₃
分子量: 197,24 g/mol
CAS号: 23996-12-5
EINECS: 245-972-9

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 107.0 to 111.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR