(1ɑ,3ɑ,5ɑ)-1,3,5-Cyclohexanetricarbonitrile ≥98.0% (通过GC)

供应商: TCI EUROPE
Danger

TCIAC1967-1GEA 0 CNY
TCIAC1967-1G
(1ɑ,3ɑ,5ɑ)-1,3,5-Cyclohexanetricarbonitrile ≥98.0% (通过GC)
(1alpha,3alpha,5alpha)-1,3,5-Cyclohexanetricarbonitrile
分子式: C₉H₉N₃
分子量: 159,19 g/mol
CAS号: 168280-46-4
UN号: 3439
ADR: 6.1,III

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 168.0 to 172.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR