4-Chloro-D-phenylalanine hydrochloride ≥98.0% (通过滴定分析)
供应商: TCI EUROPE
TCIAC2016-1GEA
0
CNY
TCIAC2016-1G
4-Chloro-D-phenylalanine hydrochloride ≥98.0% (通过滴定分析)
4-Chloro-D-phenylalanine hydrochloride
分子式:
C₉H₁₀ClNO₂ 分子量: 236,09 g/mol 溶点: 235°C |
CAS号:
147065-05-2 |
产品技术规格
Appearance | White to Light yellow powder to crystal |
Purity (Argentmetric Titration) | min. 98.0 % |
Specific rotation [a]20/D | +8.0 to +12.0 deg (C=1, H2O) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...