3-Chloro-2,2,3,3-tetrafluoropropionic acid ≥98.0% (通过滴定分析)

供应商: TCI EUROPE
Danger

TCIAC2356-5GEA 0 CNY
TCIAC2356-5G
3-Chloro-2,2,3,3-tetrafluoropropionic acid ≥98.0% (通过滴定分析)
3-Chloro-2,2,3,3-tetrafluoropropionic acid
分子式: C₃HClF₄O₂
分子量: 180,48 g/mol
沸点: 131°C
MDL编号: MFCD00153099
CAS号: 661-82-5
UN号: 3265
ADR: 8,II

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR