Copper(II) tetrafluoroborate ∼45% 在水里
供应商: TCI EUROPE
|
Danger
|
TCIAC2410-500GEA
0
CNY
TCIAC2410-500G
Copper(II) tetrafluoroborate ∼45% 在水里
Copper(II) tetrafluoroborate
分子式:
B₂CuF₈ 分子量: 237,15 g/mol |
MDL编号:
MFCD00016054 CAS号: 38465-60-0 EINECS: 253-959-4 UN号: 3264 ADR: 8,III |
产品技术规格
Appearance | Blue to Dark blue clear liquid |
Concentration (Chelometric Method) | 45.0 to 50.0 w/w% |
Water | 50.0 to 55.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...