1-Carbobenzoxy-3-methylimidazoliumtrifluoromethanesulphonate ≥98.0% (通过HPLC,总氮)

供应商: TCI EUROPE
Warning

TCIAC2420-5GEA 0 CNY
TCIAC2420-5G
1-Carbobenzoxy-3-methylimidazoliumtrifluoromethanesulphonate ≥98.0% (通过HPLC,总氮)
1-Carbobenzoxy-3-methylimidazoliumtrifluoromethanesulphonate
分子式: C₁₃H₁₃F₃N₂O₅S
分子量: 366,31 g/mol
CAS号: 163080-99-7

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR