Cinnamyl butyrate ≥97.0% (通过GC)

供应商: TCI EUROPE

TCIAC2438-25GEA 0 CNY
TCIAC2438-25G
Cinnamyl butyrate ≥97.0% (通过GC)
Cinnamyl butyrate
分子式: C₁₃H₁₆O₂
分子量: 204,27 g/mol
MDL编号: MFCD00027135
CAS号: 103-61-7
EINECS: 203-128-7

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR