(S)-(+)-1-Cbz-3-羟基吡咯烷 ≥97.0% (通过GC)

供应商: TCI EUROPE
Warning

Synonyms: (S)-1-苄氧羰基-3-羟基吡咯烷(S)-N-Z-3-Pyrrolidinol(S)-1-Carbobenzoxy-3-pyrrolidinol

TCIAC2453-1GEA 0 CNY
TCIAC2453-1G
(S)-(+)-1-Cbz-3-羟基吡咯烷 ≥97.0% (通过GC)
(S)-(+)-1-Cbz-3-羟基吡咯烷
分子式: C₁₂H₁₅NO₃
分子量: 221,26 g/mol
MDL编号: MFCD07784363
CAS号: 100858-32-0

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 74.0 to 78.0 deg-C
Specific rotation [a]20/D +21.0 to +25.0 deg (C=1, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR