2-Chloro-3-hexylthiophene ≥98.0% (通过GC)

供应商: TCI EUROPE

TCIAC2613-5GEA 1375 CNY
TCIAC2613-5G
2-Chloro-3-hexylthiophene ≥98.0% (通过GC)
2-Chloro-3-hexylthiophene
分子式: C₁₀H₁₅ClS
分子量: 202,74 g/mol
沸点: 60°C
存储条件: Refrigerator
MDL编号: MFCD18413738
CAS号: 817181-75-2

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR