(S)-2,4-Diaminobutanoic acid dihydrochloride ≥98.0% (通过滴定分析)

供应商: TCI EUROPE

TCIAD0083-1GEA 500 CNY
TCIAD0083-1G
(S)-2,4-Diaminobutanoic acid dihydrochloride ≥98.0% (通过滴定分析)
(S)-2,4-Diaminobutanoic acid dihydrochloride
分子式: C₄H₁₂Cl₂N₂O₂
分子量: 191,06 g/mol
MDL编号: MFCD00064561
CAS号: 1883-09-6
EINECS: 217-542-0

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Specific rotation [a]20/D +13.0 to +16.0 deg (C=3.6, H2O)
Solubility in Water very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR