Bromanilic acid ≥98.0% (通过滴定分析)
供应商: TCI EUROPE
|
Warning
|
TCIAD0182-10GEA
0
CNY
TCIAD0182-10G
Bromanilic acid ≥98.0% (通过滴定分析)
Bromanilic acid
分子式:
C₆H₂Br₂O₄ 分子量: 297,89 g/mol 溶点: 270°C |
MDL编号:
MFCD00058956 CAS号: 4370-59-6 |
产品技术规格
Appearance | Light yellow to Brown powder to crystal |
Purity (Iodometric Titration) | min. 98.0 % |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...