2,3-Dichloroisobutyric acid ≥98.0% (通过滴定分析)

供应商: TCI EUROPE
Danger

TCIAD0199-10GEA 1600 CNY
TCIAD0199-10G
2,3-Dichloroisobutyric acid ≥98.0% (通过滴定分析)
2,3-Dichloroisobutyric acid
分子式: C₄H₆Cl₂O₂
分子量: 156,99 g/mol
沸点: 113°C
MDL编号: MFCD00068279
CAS号: 10411-52-6
UN号: 3265
ADR: 8,III

现在订购

产品技术规格

Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 91.0 %
Purity (Neutralization titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR