3,5-Dibromosalicylaldoxime ≥98.0% (通过GC)

供应商: TCI EUROPE
Warning

TCIAD0210-5GEA 0 CNY
TCIAD0210-5G
3,5-Dibromosalicylaldoxime ≥98.0% (通过GC)
3,5-Dibromosalicylaldoxime
分子式: C₇H₅Br₂NO₂
分子量: 294,93 g/mol
MDL编号: MFCD00059607
CAS号: 21386-43-6

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 223.0 to 229.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR