Bis(6-hydroxy-2-naphthyl)disulfide ≥95.0% (通过HPLC)

供应商: TCI EUROPE
Warning

TCIAD0244-1GEA 845 CNY
TCIAD0244-1G
Bis(6-hydroxy-2-naphthyl)disulfide ≥95.0% (通过HPLC)
Bis(6-hydroxy-2-naphthyl)disulfide
分子式: C₂₀H₁₄O₂S₂
MDL编号: MFCD00004083
CAS号: 6088-51-3
EINECS: 228-025-4

现在订购

产品技术规格

Purity (HPLC) min. 95.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR