双(4-氟-3-硝基苯)砜 ≥98.0% (通过GC)

供应商: TCI EUROPE
Warning

Synonyms: 4,4′-二氟-3,3′-二硝基二苯砜4-氟-3-硝基苯砜

TCIAD0536-10GEA 0 CNY
TCIAD0536-10G
双(4-氟-3-硝基苯)砜 ≥98.0% (通过GC)
双(4-氟-3-硝基苯)砜
分子式: C₁₂H₆F₂N₂O₆S
分子量: 344,24 g/mol
MDL编号: MFCD00007057
CAS号: 312-30-1
EINECS: 206-224-7

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Solubility in Acetonitrile almost transparency
Melting point 193.0 to 197.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR