4,4'-Dinitrobiphenyl ≥99.0% (通过GC)

供应商: TCI EUROPE
Warning

TCIAD0826-5GEA 0 CNY
TCIAD0826-5G
4,4'-Dinitrobiphenyl ≥99.0% (通过GC)
4,4'-Dinitrobiphenyl
分子式: C₁₂H₈N₂O₄
分子量: 244,21 g/mol
MDL编号: MFCD00039744
CAS号: 1528-74-1
EINECS: 216-210-2

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 99.0 %
Melting point 240.0 to 244.0 deg-C
Solubility in hot Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR