7,8-Dimethylalloxazine ≥98.0% (通过滴定分析)

供应商: TCI EUROPE

TCIAD1066-100MGEA 0 CNY
TCIAD1066-100MG
7,8-Dimethylalloxazine ≥98.0% (通过滴定分析)
7,8-Dimethylalloxazine
分子式: C₁₂H₁₀N₄O₂
分子量: 242,24 g/mol
MDL编号: MFCD00005021
CAS号: 1086-80-2
EINECS: 214-120-8

现在订购

产品技术规格

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR