4,4'-Diamyloxybiphenyl ≥98.0% (通过GC)

供应商: TCI EUROPE

TCIAD1090-5GEA 0 CNY
TCIAD1090-5G
4,4'-Diamyloxybiphenyl ≥98.0% (通过GC)
4,4'-Diamyloxybiphenyl
分子式: C₂₂H₃₀O₂
分子量: 326,48 g/mol
MDL编号: MFCD00059432
CAS号: 21470-41-7

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 134.0 to 138.0 deg-C
Solubility in hot Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR