4,4'-Dibutoxyazoxybenzene

供应商: TCI EUROPE

TCIAD1096-1GEA 3150 CNY
TCIAD1096-1G
4,4'-Dibutoxyazoxybenzene
4,4'-Dibutoxyazoxybenzene
分子式: C₂₀H₂₆N₂O₃
MDL编号: MFCD00048767
CAS号: 17051-01-3

现在订购

产品技术规格

Appearance Light yellow to Brown powder to crystal

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR