9-(4'-Dimethylaminophenyl)-2,6,7-trihydroxyfluorone sulphate hydrate ≥96.0% (通过HPLC)
供应商: TCI EUROPE
TCIAD1118-1GEA
0
CNY
TCIAD1118-1G
9-(4'-Dimethylaminophenyl)-2,6,7-trihydroxyfluorone sulphate hydrate ≥96.0% (通过HPLC)
9-(4'-Dimethylaminophenyl)-2,6,7-trihydroxyfluorone sulphate hydrate
分子式:
C₂₁H₁₇NO₅ |
MDL编号:
MFCD00035151 CAS号: 71501-19-4 EINECS: 275-549-4 |
产品技术规格
Appearance | Orange to Brown powder to crystal |
Purity (HPLC) | min. 96.0 area% |
Water | max. 15.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...