4-羟基-2,5-二甲基l-3(2H)-呋喃酮 15% ∼1.2 mol/L 在丙二醇中

供应商: TCI EUROPE
Warning

Synonyms: 天然呋喃酮2,5-二甲基-4-羟基-3(2H)-呋喃酮草莓呋喃酮

TCIAD1884-25MLEA 810 CNY
TCIAD1884-25ML
4-羟基-2,5-二甲基l-3(2H)-呋喃酮 15% ∼1.2 mol/L 在丙二醇中
4-羟基-2,5-二甲基l-3(2H)-呋喃酮
分子式: C₆H₈O₃
分子量: 128,13 g/mol
MDL编号: MFCD00010706
CAS号: 3658-77-3
EINECS: 222-908-8

现在订购

产品技术规格

Appearance White to Yellow to Green clear liquid
Concentration (Neutralization titration) 15.0 to 16.0 w/w%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR