1,4-Diisopropenylbenzene ≥98.0% (通过GC)

供应商: TCI EUROPE

TCIAD1973-10GEA 0 CNY
TCIAD1973-10G
1,4-Diisopropenylbenzene ≥98.0% (通过GC)
1,4-Diisopropenylbenzene
分子式: C₁₂H₁₄
分子量: 158,24 g/mol
存储条件: Refrigerator
MDL编号: MFCD00043637
CAS号: 1605-18-1
EINECS: 216-515-0

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 63.0 to 67.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR