1,5,9-Decatriene ≥95.0% (通过GC)
供应商: TCI EUROPE
|
Danger
|
TCIAD1993-25MLEA
0
CNY
TCIAD1993-25ML
TCIAD1993-5ML
1,5,9-Decatriene ≥95.0% (通过GC)
1,5,9-Decatriene
分子式:
C₁₀H₁₆ 沸点: 55°C 闪点: 46°C |
MDL编号:
MFCD00055625 CAS号: 13393-64-1 EINECS: 236-474-2 UN号: 3295 ADR: 3,III |
产品技术规格
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...