2,3-Dichloropropionamide ≥98.0% (通过GC)

供应商: TCI EUROPE

TCIAD1999-10GEA 0 CNY
TCIAD1999-10G
2,3-Dichloropropionamide ≥98.0% (通过GC)
2,3-Dichloropropionamide
分子式: C₃H₅Cl₂NO
分子量: 141,98 g/mol
MDL编号: MFCD00040311
CAS号: 19433-84-2
EINECS: 243-056-3

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 106.0 to 110.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR