1,2-Diacetoxybenzene ≥98.0% (通过GC)
供应商: TCI EUROPE
|
Warning
|
TCIAD2044-25GEA
0
CNY
TCIAD2044-25G
1,2-Diacetoxybenzene ≥98.0% (通过GC)
1,2-Diacetoxybenzene
分子式:
C₁₀H₁₀O₄ 分子量: 194,19 g/mol 沸点: 140°C |
MDL编号:
MFCD00017219 CAS号: 635-67-6 EINECS: 211-240-2 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 62.0 to 65.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...