反式-1,4-二噁烷-2,3-二醇 ≥96.0% (通过GC)
供应商: TCI EUROPE
Synonyms:
反式-2,3-二羟基-1,4-二氧杂环己烷
, 乙二醛单乙二醇缩醛
TCIAD2066-10GEA
600
CNY
TCIAD2066-10G
反式-1,4-二噁烷-2,3-二醇 ≥96.0% (通过GC)
反式-1,4-二噁烷-2,3-二醇
分子式:
C₄H₈O₄ 分子量: 120,10 g/mol |
MDL编号:
MFCD00006572 CAS号: 4845-50-5 EINECS: 225-431-3 |
产品技术规格
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...