(β,β-Dimethylacryl)shikonin ≥90.0% (通过HPLC)

供应商: TCI EUROPE

TCIAD2118-100MGEA 0 CNY
TCIAD2118-100MG
(β,β-Dimethylacryl)shikonin ≥90.0% (通过HPLC)
(beta,beta-Dimethylacryl)shikonin
分子式: C₂₁H₂₂O₆
分子量: 370,40 g/mol
MDL编号: MFCD00143687
CAS号: 24502-79-2

现在订购

产品技术规格

Appearance Dark red to Brown powder to crystal
Purity (HPLC) min. 90.0 area%
Melting point 103.0 to 105.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR