2,5-Di-tert-amylbenzoquinone ≥95.0% (通过GC)

供应商: TCI EUROPE

TCIAD2152-5GEA 870 CNY
TCIAD2152-5G
2,5-Di-tert-amylbenzoquinone ≥95.0% (通过GC)
2,5-Di-tert-amylbenzoquinone
分子式: C₁₆H₂₄O₂
MDL编号: MFCD00142843
CAS号: 4584-63-8

现在订购

产品技术规格

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 95.0 %
Melting point 135.0 to 139.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR