(+)-1,4-O-二苯基-D-苏醇 ≥98.0% (通过GC)

供应商: TCI EUROPE

Synonyms: (+)-(2R,3R)-1,4-双(苄氧基)-2,3-丁二醇(+)-1,4-二-O-苄基-D-苏糖醇1,4-二-O-苄基-D-苏醇

TCIAD2239-1GEA 1825 CNY
TCIAD2239-1G
(+)-1,4-O-二苯基-D-苏醇 ≥98.0% (通过GC)
(+)-1,4-O-二苯基-D-苏醇
分子式: C₁₈H₂₂O₄
分子量: 302,37 g/mol
MDL编号: MFCD00077722
CAS号: 91604-41-0

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Optical purity (LC) min. 98.0 ee%
Melting point 56.0 to 59.0 deg-C
Specific rotation [a]20/D +6.0 to +6.5 deg (C=5, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR