4'-(Dimethylamino)benzylidene-4-nitroaniline

供应商: TCI EUROPE

TCIAD2264-5GEA 755 CNY
TCIAD2264-5G
4'-(Dimethylamino)benzylidene-4-nitroaniline
4'-(Dimethylamino)benzylidene-4-nitroaniline
分子式: C₁₅H₁₅N₃O₂
MDL编号: MFCD00086550
CAS号: 896-05-9

现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR