4-Dibenzyl 1-naphthyl ketone ≥98.0% (通过HPLC)

供应商: TCI EUROPE

TCIAD2277-1GEA 0 CNY
TCIAD2277-1G
4-Dibenzyl 1-naphthyl ketone ≥98.0% (通过HPLC)
4-Dibenzyl 1-naphthyl ketone
分子式: C₂₅H₂₀O
分子量: 336,43 g/mol
MDL编号: MFCD00191634
CAS号: 158098-50-1

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 83.0 to 87.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR