4-Diazo-3-methoxydiphenylamine sulfate ≥96.0% (通过HPLC,滴定分析)

供应商: TCI EUROPE
Warning

TCIAD2285-25GEA 585 CNY
TCIAD2285-25G
4-Diazo-3-methoxydiphenylamine sulfate ≥96.0% (通过HPLC,滴定分析)
4-Diazo-3-methoxydiphenylamine sulfate
分子式: C₁₃H₁₂N₃O
MDL编号: MFCD00083004
CAS号: 36305-05-2
EINECS: 252-959-1

现在订购

产品技术规格

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Coupling titration) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR