2-(4-Nitrophenyl)-4,4,5,5-tetramethylimidazoline-3-oxide-1-oxyl free radical

供应商: TCI EUROPE

TCIAD2295-100MGEA 0 CNY
TCIAD2295-100MG
2-(4-Nitrophenyl)-4,4,5,5-tetramethylimidazoline-3-oxide-1-oxyl free radical
2-(4-Nitrophenyl)-4,4,5,5-tetramethylimidazoline-3-oxide-1-oxyl
分子式: C₁₃H₁₆N₃O₄
分子量: 278,29 g/mol
MDL编号: MFCD00191637
CAS号: 38582-73-9

现在订购

产品技术规格

Appearance Green to Dark green powder to crystal

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR