1,3-Dimethyl-2-phenyl-1,3,2-diazaphospholidine ≥92.0% (通过滴定分析)

供应商: TCI EUROPE

TCIAD2300-5GEA 0 CNY
TCIAD2300-5G
1,3-Dimethyl-2-phenyl-1,3,2-diazaphospholidine ≥92.0% (通过滴定分析)
1,3-Dimethyl-2-phenyl-1,3,2-diazaphospholidine
分子式: C₁₀H₁₅N₂P
分子量: 194,22 g/mol
闪点: 108°C
存储条件: Refrigerator
MDL编号: MFCD00043134
CAS号: 22429-12-5

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (Nonaqueous Titration) min. 92.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR