2,2-Dibromo-1,3-diphenyl-1,3-propanedione ≥98.0% (通过滴定分析)
供应商: TCI EUROPE
TCIAD2324-10GEA
0
CNY
TCIAD2324-10G
2,2-Dibromo-1,3-diphenyl-1,3-propanedione ≥98.0% (通过滴定分析)
2,2-Dibromo-1,3-diphenyl-1,3-propanedione
分子式:
C₁₅H₁₀Br₂O₂ 分子量: 382,05 g/mol |
MDL编号:
MFCD00068642 CAS号: 16619-55-9 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...