2,5-Dicyanofuran ≥98.0% (通过GC)

供应商: TCI EUROPE
Danger

TCIAD2416-100MGEA 0 CNY
TCIAD2416-100MG
2,5-Dicyanofuran ≥98.0% (通过GC)
2,5-Dicyanofuran
分子式: C₆H₂N₂O
分子量: 118,10 g/mol
MDL编号: MFCD00671519
CAS号: 58491-62-6
EINECS: 261-287-8
UN号: 3439
ADR: 6.1,III

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 63.0 to 66.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR