2,3-Dicyano-1,4-dihydroxy-5-nitronaphthalene ≥98.0% (通过HPLC,总氮)

供应商: TCI EUROPE
Danger

TCIAD2434-5GEA 0 CNY
TCIAD2434-5G
2,3-Dicyano-1,4-dihydroxy-5-nitronaphthalene ≥98.0% (通过HPLC,总氮)
2,3-Dicyano-1,4-dihydroxy-5-nitronaphthalene
分子式: C₁₂H₅N₃O₄
分子量: 255,19 g/mol
MDL编号: MFCD00671523
CAS号: 4655-62-3
UN号: 3439
ADR: 6.1,III

现在订购

产品技术规格

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR