4-Dimethylamino-4'-nitrostilbene ≥98.0% (通过HPLC,滴定分析)

供应商: TCI EUROPE
Danger

TCIAD2505-5GEA 0 CNY
TCIAD2505-5G
4-Dimethylamino-4'-nitrostilbene ≥98.0% (通过HPLC,滴定分析)
4-Dimethylamino-4'-nitrostilbene
分子式: C₁₆H₁₆N₂O₂
MDL编号: MFCD00041877
CAS号: 4584-57-0
UN号: 2811
ADR: 6.1,III

现在订购

产品技术规格

Appearance White to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR