(-)-(R)-N,N-二甲基-1-[(S)-1′,2-双(二苯基膦基)二茂铁基]乙胺 ≥98.0% (通过GC)

供应商: TCI EUROPE
Warning

Synonyms: [R,R]-1-[1-二甲基氨乙基]-1′,2-双[二苯基膦]二茂铁

TCIAD2537-100MGEA 0 CNY
TCIAD2537-100MG
(-)-(R)-N,N-二甲基-1-[(S)-1′,2-双(二苯基膦基)二茂铁基]乙胺 ≥98.0% (通过GC)
(-)-(R)-N,N-二甲基-1-[(S)-1′,2-双(二苯基膦基)二茂铁基]乙胺
分子式: C₃₈H₃₇FeNP₂
分子量: 625,51 g/mol
MDL编号: MFCD00075284
CAS号: 74311-56-1

现在订购

产品技术规格

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Optical purity (HPLC) min. 98.0 ee%
Melting point 138.0 to 144.0 deg-C
Specific rotation [a]20/D -330.0 to -360.0 deg (C=0.5, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR