4-(2,4-Dinitrobenzyl)pyridine

供应商: TCI EUROPE

TCIAD2543-1GEA 1025 CNY
TCIAD2543-1G
4-(2,4-Dinitrobenzyl)pyridine
4-(2,4-Dinitrobenzyl)pyridine
分子式: C₁₂H₉N₃O₄
MDL编号: MFCD01321164
CAS号: 1603-85-6

现在订购

产品技术规格

Appearance Gray to Dark purple to Black powder to crystal
Purity (GC) min. 95.0 %
Melting point 77.0 to 80.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR