(1S,4S)-(+)-2,5-二氮双环[2.2.1]庚烷 ≥98.0% (通过滴定分析)
供应商: TCI EUROPE
|
Warning
|
Synonyms:
(1S,4S)-2,5-二氮双环[2.2.1]庚烷
TCIAD2658-1GEA
0
CNY
TCIAD2658-1G
(1S,4S)-(+)-2,5-二氮双环[2.2.1]庚烷 ≥98.0% (通过滴定分析)
(1S,4S)-(+)-2,5-二氮双环[2.2.1]庚烷
分子式:
C₅H₁₀N₂ 分子量: 259,97 g/mol |
MDL编号:
MFCD01321289 CAS号: 132747-20-7 |
产品技术规格
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (Argentmetric Titration) | min. 98.0 % |
Specific rotation [a]20/D | +18.0 to +23.0 deg (C=1,H2O) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...