2,2-Dichlorocyclopropyl phenyl sulphide ≥97.0% (通过GC)

供应商: TCI EUROPE

TCIAD2660-1GEA 0 CNY
TCIAD2660-1G
2,2-Dichlorocyclopropyl phenyl sulphide ≥97.0% (通过GC)
2,2-Dichlorocyclopropyl phenyl sulphide
分子式: C₉H₈Cl₂S
分子量: 219,12 g/mol
MDL编号: MFCD00800592
CAS号: 63289-85-0

现在订购

产品技术规格

Appearance Light yellow to Brown to Dark green powder to lump
Purity (GC) min. 97.0 %
Freezing point 22.0 to 26.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR