2,2-Diisoamyl-1,3-propanediol ≥98.0% (通过GC)

供应商: TCI EUROPE

TCIAD2800-5GEA 0 CNY
TCIAD2800-5G
2,2-Diisoamyl-1,3-propanediol ≥98.0% (通过GC)
2,2-Diisoamyl-1,3-propanediol
分子式: C₁₃H₂₈O₂
分子量: 216,37 g/mol
MDL编号: MFCD03701099
CAS号: 403519-64-2

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 91.0 to 95.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR