3-Dibutylamino-1-propyne ≥98.0% (通过滴定分析)
供应商: TCI EUROPE
|
Warning
|
TCIAD2817-5MLEA
0
CNY
TCIAD2817-5ML
3-Dibutylamino-1-propyne ≥98.0% (通过滴定分析)
3-Dibutylamino-1-propyne
分子式:
C₁₁H₂₁N 分子量: 167,30 g/mol |
MDL编号:
MFCD00041673 CAS号: 6336-58-9 |
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...