4-(二氰基亚甲基)-2-甲基-6-(4-二甲基氨基苯乙烯基)-4H-吡喃 ≥95.0% (通过HPLC)

供应商: TCI EUROPE
Warning

Synonyms: DCM

TCIAD2849-500MGEA 0 CNY
TCIAD2849-500MG
4-(二氰基亚甲基)-2-甲基-6-(4-二甲基氨基苯乙烯基)-4H-吡喃 ≥95.0% (通过HPLC)
4-(二氰基亚甲基)-2-甲基-6-(4-二甲基氨基苯乙烯基)-4H-吡喃
分子式: C₁₉H₁₇N₃O
分子量: 303,37 g/mol
MDL编号: MFCD00051341
CAS号: 51325-91-8
EINECS: 257-137-6
UN号: 1325
ADR: 4.1,III

现在订购

产品技术规格

Appearance Dark red to Brown powder to crystal
Purity (HPLC) min. 95.0 area%
Melting point 217.0 to 223.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR