5,15-Diphenylporphyrin ≥90.0% (通过HPLC)

供应商: TCI EUROPE

TCIAD2860-100MGEA 0 CNY
TCIAD2860-100MG
5,15-Diphenylporphyrin ≥90.0% (通过HPLC)
5,15-Diphenylporphyrin
分子式: C₃₂H₂₂N₄
分子量: 462,56 g/mol
MDL编号: MFCD08276335
CAS号: 22112-89-6

现在订购

产品技术规格

Appearance Dark red to Dark purple to Dark blue powder to crystal
Purity (HPLC) min. 90.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR