1,3-Dichloro-2,4,6-trifluorobenzene ≥97.0% (通过GC)

供应商: TCI EUROPE
Warning

TCIAD2868-5GEA 1175 CNY
TCIAD2868-5G
1,3-Dichloro-2,4,6-trifluorobenzene ≥97.0% (通过GC)
1,3-Dichloro-2,4,6-trifluorobenzene
分子式: C₆HCl₂F₃
MDL编号: MFCD00012242
CAS号: 2368-53-8

现在订购

产品技术规格

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %
Specific gravity (20/20) 1.5970 to 1.6010

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR